Thermo Scientific - TSX2 HP Assure Laboratory Freezer - TSX2-2520FV - Technical Data Sheet
Technical specifications for the Thermo Scientific TSX2 HP Assure Manual Defrost Laboratory Freezer, model TSX2-2520FV, featuring a solid door and 24.7 cu. ft. (700 L) capacity for laboratory storage at -25C to -15C. Operates on 220-240V, 50/60Hz, single-phase power at 1.5A. Features -20C setpoint, touchscreen control, manual defrost, R290 refrigerant, adjustable temperature alarms, power failure alarm, 24-hour/7-day temperature history, RS485 connectivity, USB data export, and remote alarm cont
Description:
Attached is the technical data sheet also known as a specification sheet for model TSX2-2520FV.
Environment:
TSX2 HP Assure, TSX2 Assure Series, TSX2 Assure High Performance Freezer, TSX2-2520FV, specs, tds
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific - TSX2 HP Assure Laboratory Freezer - TSX2-2520FV - Technical Data Sheet.pdf | October 23, 2026 |