Thermo Fisher Scientific
Thermo Scientific - General Purpose Undercounter Lab Refrigerator - TSU405GA Technical Data Sheet
This article provides a technical overview of the Thermo Scientific TSU405GA glass door undercounter laboratory refrigerator, detailing purpose, capacity, temperature control, power, and certifications. It lists 4.5 cu ft storage, +2°C to +15°C setpoint, +5°C factory setting, R290 refrigerant, forced-air condenser, alarms, data logging, LCD controller, 115 V/60 Hz NEMA 5-15P, indoor use, cETLus, ENERGY STAR, NSF/ANSI 456, and cabinet dimensions.
Description: 
The attached is the TSU405GA technical data sheet which provides specifications and technical data on this model.     
Environment:
TSU405GA, TDS, technical data sheet, tsu405GA specs, tsu405GA technical data sheet
  • Was this article helpful?