Thermo Scientific - TSX1 HP Flex Laboratory Refrigerator - TSX1-0505SA - Technical Data Sheet
Technical specifications for the Thermo Scientific TSX1 HP Flex Laboratory Refrigerator, model TSX1-0505SA, featuring a solid door and 4.6 cu. ft. (130 L) capacity for laboratory storage at 2C to 8C. Operates on 115V, 60Hz, single-phase power at 2.2A. Features digital temperature control, automatic defrost, R600 refrigerant, adjustable temperature alarms, power failure alarm, RS485 connectivity, USB data export, and remote alarm contacts.
Description:
Attached is the technical data sheet also known as a specification sheet for model TSX1-0505SA.
Environment:
TSX1 HP Flex, TSX1 Flex Series, TSX1 Flex High Performance Refrigerator, TSX1-0505SA, specs, tds
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific - TSX1 HP Flex Laboratory Refrigerator - TSX1-0505SA - Technical Data Sheet.pdf | October 22, 2026 |