Thermo Scientific - TSX2 HP Blood Bank Refrigerator - TSX2-2204BA - Technical Data Sheet
Technical specifications for the Thermo Scientific TSX2 Series Blood Bank Refrigerator, model TSX2-2204BA, featuring 22.2 cu. ft. (629 L) capacity and six adjustable drawers for blood storage at 3C to 5C. Operates on 115V, 60Hz, single-phase power at 2.3A. Features 4C setpoint, digital temperature control, automatic defrost, R600 refrigerant, adjustable temperature alarms, power failure alarm, RTD sensor, RS485 connectivity, USB data export, remote alarm contacts, and cULus/FDA listings.
Description:
Attached is the technical data sheet also known as a specification sheet for model TSX2-2204BA.
Environment:
TSX2 HP Assure, TSX2 Assure Series, TSX2 Assure High Performance Refrigerator, TSX2-2204BA, specs, tds, blood bank refrigerator, TSX2 BBR
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Scientific - TSX2 HP Blood Bank Refrigerator - TSX2-2204BA - Technical Data Sheet.pdf | October 26, 2026 |