Thermo Scientific General Purpose Undercounter Lab Refrigerator - TSU405SA Technical Data Sheet
This technical data sheet summarizes the Thermo Scientific TSU405SA undercounter laboratory refrigerator, covering capacity, temperature control, electrical specs, certifications, dimensions, and key features for lab storage. Specs include 4.5 cu ft capacity, +2°C to +15°C setpoint, R290 refrigerant, forced-air condenser, alarms, data logging, LCD controller, 115 V/60 Hz NEMA 5-15P, ENERGY STAR, NSF/ANSI 456, cETLus, indoor use, and performance map.
Description:
The attached document is the technical data sheet for the TSU405SA which provides a variety of specification or technical data.
Environment:
TSU405SA, technical data sheet, specifications, TSU405SA TDS
Attachment(s):
| File | Last Modified |
|---|---|
| TSU405SA Technical Data Sheet - Rev A.pdf | March 25, 2026 |