Thermo Fisher Scientific
157-3000-00 - Rev E - EC-105 Electrophoresis Power Supply - User Manual
Description: 
User manual
Environment:
EC-105 Electrophoresis Power Supply
The EC-105 Power Supply is intended for use with any electrophoretic device designed to operate below 250 Volts and 500 milliAmps DC.
  • Was this article helpful?