7007309 Rev 1 - Horizontal Systems Models B1A_B1_B2 and B3 - Operating and Maintenance Manual
The Operating and Maintenance Manual provides comprehensive guidance on the operation and maintenance of Horizontal Mini gel Systems (Models B1A, B1, B2) and the Self Recirculation System (Model B3). It covers specifications, recommended running conditions, and safety instructions. The manual emphasizes hassle-free gel casting, precise photo documentation, and a built-in recirculation system to prevent pH and ionic gradients, ensuring high-resolution, reproducible results.
Description:
Operating and maintenance manual
Environment:
Horizontal Minigel Systems - Models:
- B1A
- B1
- B2
Self Recirculation System - Model:
- B3
Specifications and recommended running conditions:
| Model | B1A | B1 | B2 | B3 |
| Gel Size (Wx L in cm.) | 7x8 | 9x11 | 12x14 | 12x14 |
| Buffer Capacity | 400ml | 600ml | 800ml | 1000ml |
| Voltage Requirements (V) | 20-150 | 20-150 | 20-150 | 20-150 |
| Time Requirements (minutes) | 30-60 | 45-90 | 60-120 | 60-120 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007309 - Rev 1 - Horizontal Systems, Models B1A, B1, B2 and B3 - Operating and Maintenance Manual.pdf | December 15, 2021 |