Thermo Fisher Scientific
7007320 - Rev 2 - Horizontal Electrophoresis System, Model D2 and D3-14 - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
  • Owl Model D3-14 Horizontal Agarose Gel Electrophoresis System
  • Owl Model D2 Horizontal Agarose Gel Electrophoresis System
Recommended running conditions:
Model D3-14 D2
Gel Size (W x L in cm) 23 x 14 14.4 x 10.2
Buffer Capacity (ml) 800ml 600ml
Voltage Requirements (V) 20-150 20-150
Time Requirements (min) 30-60 30-60