7007320 - Rev 2 - Horizontal Electrophoresis System Model D2 and D3-14 - Operating and Maintenance Manual
The D2 and D3-14 Operating and Maintenance Manual provides comprehensive guidelines on the operation and maintenance of the Owl Model D2 and D3-14 Horizontal Agarose Gel Electrophoresis Systems. It covers recommended running conditions, safety information, system components, and detailed procedures for using the system, running gels, and maintenance. The manual emphasizes the importance of following safety protocols to avoid equipment damage and personal injury.
Description:
Operating and maintenance manual
Environment:
- Owl Model D3-14 Horizontal Agarose Gel Electrophoresis System
- Owl Model D2 Horizontal Agarose Gel Electrophoresis System
Recommended running conditions:
| Model | D3-14 | D2 |
| Gel Size (W x L in cm) | 23 x 14 | 14.4 x 10.2 |
| Buffer Capacity (ml) | 800ml | 600ml |
| Voltage Requirements (V) | 20-150 | 20-150 |
| Time Requirements (min) | 30-60 | 30-60 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007320 - Rev 2 - Horizontal Electrophoresis System, Model D2 and D3-14 - Operating and Maintenance Manual.pdf | December 15, 2021 |