7007325 - Rev 1 - Horizontal Electrophoresis System Model D4 - Operating and Maintenance Manual
The document titled "7007325 - Rev 1 - Horizontal Electrophoresis System Model D4 - Operating and Maintenance Manual" provides comprehensive information on operating and maintaining the Owl Horizontal Agarose Gel Electrophoresis System, Model D4. It includes specifications, safety guidelines, setup instructions, and troubleshooting tips. Key features include dual gel trays, UVT gel trays, and a stand-alone casting platform. It emphasizes electrical safety and proper use to avoid damage.
Description:
Operating and maintenance manual
Environment:
Owl Horizontal Agarose Gel Electrophoresis System, model:
- D4
Specifications and recommended running conditions:
| Gel size (W x L cm) | 16 x 17 | |
| Buffer capacity | 800 | 1 tray |
| 1600 | 2 trays | |
| Voltage requirements (V) | 100V minimum | |
| Time requirements | 4½ hours |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007325 - Rev 1 - Horizontal Electrophoresis System, Model D4 - Operating and Maintenance Manual.pdf | December 15, 2021 |