Thermo Fisher Scientific
7007332 - Rev 0 - Semidry Electroblotter, Models HEP-1 and HEP-3 - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Thermo Scientific Semidry Electroblotter, models:
  • HEP-1
  • HEP-3
Item Description Catalog No.
Complete System HEP-1
Transfer Area 20cm W x 20cm L
Footprint 25cm W x 25cm D x 5cm H
Complete System HEP-3
Transfer Area 35cm W x 45cm L
Footprint 40cm W x 51cm D x 5cm H