7007332 Rev 0 - Semidry Electroblotter Models HEP-1 and HEP-3 - Operating and Maintenance Manual
The HEP-1 and HEP-3 manual provides comprehensive guidelines for the operation and maintenance of the HEP-1 and HEP-3 Semidry Electroblotter models. The document includes important safety instructions, product descriptions, setup procedures, troubleshooting tips, and proper handling techniques for various components such as filter papers, membranes, and transfer buffers. It emphasizes the importance of following the manual to ensure safety and optimal performance of the equipment.
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Semidry Electroblotter, models:
- HEP-1
- HEP-3
| Item Description | Catalog No. |
| Complete System | HEP-1 |
| Transfer Area | 20cm W x 20cm L |
| Footprint | 25cm W x 25cm D x 5cm H |
| Complete System | HEP-3 |
| Transfer Area | 35cm W x 45cm L |
| Footprint | 40cm W x 51cm D x 5cm H |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007332 - Rev 0 - Semidry Electroblotter, Models HEP-1 and HEP-3 - Operating and Maintenance Manual.pdf | December 15, 2021 |