7007335 Rev 1 - Water Cooled - Dual Gel Electrophoresis System Models P8DS_P9DS and P10DS - Operating and Maintenance Manual
The Water Cooled, Dual Gel Electrophoresis System, Models P8DS, P9DS and P10DS - Operating and Maintenance Manual" provides comprehensive information on the operation and maintenance of the specified electrophoresis systems. It includes specifications for gel sizes, buffer chamber capacities, running conditions, and time requirements. The manual details the steps for operating the system, including setting up, running, and finishing procedures, along with important safety notes.
Description:
Operating and maintenance manual
Environment:
Water Cooled, Dual Gel Electrophoresis System, Models:
- P8DS
- P9DS
- P10DS
Specifications:
| Unit/Model Number | P8DS | P9DS | P10DS |
| Gel size |
10cmW x 10cmL 10cmW x 8cmL |
16cmW x 14cmL 16cmW x 16cmL |
20cmW x 20cmL |
| Upper Buffer Chamber Capacity | 170ml | 400ml | 600ml |
| Lower Buffer Chamber Capacity | 240ml | 300ml | 800ml |
| Total Running Buffer | 250ml | 650ml | 1250ml |
| Total Buffer Capacity | 250ml | 650ml | 1250ml |
| Current, Constant | 15-35mA/gel | 15-50mA/gel | 15-75mA/gel |
| Time Requirements | 30-90 min. | 60-120 min. | 60-180 min. |
| Sample Capacity | 20 | 30 | 40 |
| Dimensions (cm) H x Wx D | 15 x 17 x 12 | 21 x 24 x 14 | 24 x 30.5 x 16.5 |
| Glass Size (cm) Wx L | 10 x 10 | 16 x 14 | 20 x 20 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007335 - Rev 1 - Water Cooled, Dual Gel Electrophoresis System, Models P8DS, P9DS and P10DS - Operating and Maintenance Manual.pdf | January 19, 2022 |