Thermo Fisher Scientific
7007341 - Rev 1 - Single Sided Vertical Electrophoresis System, P81 - Operating and Maintenance Manual
Description: 
Operating and maintenance manual 
Environment:
Single Sided Vertical Electrophoresis System, P81, specifications:
Unit/Model Number P81
Gel size 10cmW x 10cmL and 10cmW x 8cmL
Upper Buffer Chamber Capacity 100ml
Lower Buffer Chamber Capacity 50ml
Total Running Buffer 150ml
Total Buffer Capacity 150ml
Current, Constant 15-35mA/gel
Time Requirements 30-90 min.
Sample Capacity 10
Dimensions (cm) H x W x D 12.7 x 18.4 x 8.3
Glass Size (cm)W x L 10 x 10