7007341 Rev 1 - Single Sided Vertical Electrophoresis System P81 - Operating and Maintenance Manual
The System P81 manual" provides comprehensive information on the operation and maintenance of the Single Sided Vertical Electrophoresis System, model P81. It includes specifications, safety instructions, parts list, optional equipment, and detailed procedures for using and maintaining the system. Key specifications include a gel size of 10cmW x 10cmL, buffer chamber capacities, and current requirements. The manual emphasizes the importance of following safety guidelines and using the equipment a
Description:
Operating and maintenance manual
Environment:
Single Sided Vertical Electrophoresis System, P81, specifications:
| Unit/Model | Number P81 |
| Gel size | 10cmW x 10cmL and 10cmW x 8cmL |
| Upper Buffer Chamber Capacity | 100ml |
| Lower Buffer Chamber Capacity | 50ml |
| Total Running Buffer | 150ml |
| Total Buffer Capacity | 150ml |
| Current, Constant | 15-35mA/gel |
| Time Requirements | 30-90 min. |
| Sample Capacity | 10 |
| Dimensions (cm) H x W x D | 12.7 x 18.4 x 8.3 |
| Glass Size (cm)W x L | 10 x 10 |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007341 - Rev 1 - Single Sided Vertical Electrophoresis System, P81 - Operating and Maintenance Manual.pdf | December 15, 2021 |