7007359 Rev 0 - Tank Electroblotting System Model VEP-2 XM - Installation and Operation Manual
The Tank Electroblotting System Model VEP-2 XM Installation and Operation Manual (7007359 Rev 0) provides comprehensive guidelines on setting up and operating the VEP-2 XM system. It includes specifications such as dimensions, buffer requirements, gel size, and current requirements. The manual details the setup process, running conditions, and safety information, emphasizing the importance of following instructions to avoid damage and ensure optimal performance.
Description:
Installation and operation manual
Environment:
Tank Electroblotting System, Model VEP-2 XM, specifications and recommended running conditions:
| Model VEP-2 | |
| Dimensions (Footprint): |
5.125in W x 6.44in H x 7.3in D (13cm W x 16.3cm H x 18.5cm D) |
| Buffer required: | 1300ml |
| Gel size(maximum): | 10cm x 10cm |
| Current requirements: | 200-500mA |
| Time required: | 45 minutes to 2 hours |
Attachment(s):
| File | Last Modified |
|---|---|
| 7007359 - Rev 0 - Tank Electroblotting System, Model VEP-2 XM - Installation and Operation Manual.pdf | December 15, 2021 |