7217584 Rev 0 - Electrophoresis Power Supply EC3000XL - Operating and Maintenance Manual
The Electrophoresis Power Supply EC3000XL Manual provides detailed information on the operation and maintenance of the EC3000XL Electrophoresis Power Supply. It includes safety instructions, features, and operation modes (constant voltage, current, power, or temperature). The manual also covers maintenance procedures such as cleaning, fuse replacement, and checking electrical components, as well as listing available accessories and replacement parts.
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Electrophoresis Power Supply, EC3000XL
AC Input Power
- 95-265VAC, 50-60Hz, 500VA Max
Environmental
- Operating temperature: 0-40°C, 0-95% R.H. non-condensing
- Altitude: 2000m
- Overvoltage category II, IEC664
- Pollution degree 2, IEC664
DC Output Power
- 20-3000VDC, 400W Max
- 1-400mA, 400W Max
- Ripple: ±1%
- Drift: ±1%, after 30-minute warm-up
Attachment(s):
| File | Last Modified |
|---|---|
| 7217584 - Rev 0 - Electrophoresis Power Supply, EC3000XL - Operating and Maintenance Manual.pdf | December 15, 2021 |