7217692 Rev 0 - Gel Caster Models JGC-3_JGC-2 and JGC-4 - Operating and Maintenance Manual
The document "7217692 Rev 0 - Gel Caster Models JGC-3, JGC-2, and JGC-4 - Operating and Maintenance Manual" provides detailed instructions on the operation and maintenance of these gel caster models. It describes the patented Gel Casting System, which allows for casting polyacrylamide gels without taping or sealing the plates. The manual includes specifications, parts lists, setup instructions, and care guidelines for each model, ensuring efficient and accurate gel casting.
Description:
Operating and maintenance manual. This patented Gel Casting System is a unique method to cast 1 to 4 polyacrylamide gels without taping or special sealing of the gel plates.
Environment:
Gel Caster Models:
- JGC-3
- JGC-2
- JGC-4
Specifications:
| Unit/Model Number | JGC-4 | JGC-2 | JGC-3 |
| Gel size |
10cmW x 10cmL 10cmW x 8cmL |
16cmW x 14cmL 16cmW x 16cmL |
20cmW x 20cmL |
| Footprint (cm) W x D | 13 x 13 | 17 x 18 | 17 x 23 |
| Glass Size (cm) W x L |
10cmW x 10cmL 10cmW x 8cmL |
16cmW x 14cmL 16cmW x 16cmL |
20cmW x 20cmL |
| Approximate Gel Volume | 6ml | 15ml | 28ml |
| Compatible with Unit | P8DS, P81, P82 | P9DS | P10DS |
Attachment(s):
| File | Last Modified |
|---|---|
| 7217692 - Rev 0 - Gel Caster, Models JGC-3, JGC-2, and JGC-4 - Operating and Maintenance Manual.pdf | December 15, 2021 |