Thermo Fisher Scientific
7217837 - Rev 0 - Multiple Gradient Caster, Models P7-CST, P1-CST and P2-CST - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Multiple Gradient Caster, models:
  • P7-CST
  • P1-CST
  • P2-CST
Specifications:
Unit/Model Number P7-CST P1-CST P2-CST
Gel size
10cm W x 10cm L
10cm W x 8cm L
16cm W x 14cm L
16cm W x 16cm L
20cm W x 20cm L
Sample Capacity 20 30 40
Dimensions (cm) W x D x H 15 x 13 x 13 20 x 13 x 18 25 x 13 x 23
Glass Size(cm)WxL 10 x 10 16 x 14 20 x 20
Compatible w/ Owl Units P8DS, P82 P9DS P10DS