7217837 Rev 0 - Multiple Gradient Caster Models P7-CST_P1-CST and P2-CST - Operating and Maintenance Manual
The Multiple Gradient Caster Models P7-CST, P1-CST, and P2-CST - Operating and Maintenance Manual" provides detailed instructions on the operation and maintenance of these gradient casters. It includes important notices, safety instructions, assembly guidelines, and specifications for each model. The manual emphasizes proper use, handling of electrostatic discharge-sensitive parts, and the importance of qualified personnel for internal adjustments
Description:
Operating and maintenance manual
Environment:
Multiple Gradient Caster, models:
- P7-CST
- P1-CST
- P2-CST
Specifications:
| Unit/Model Number | P7-CST | P1-CST | P2-CST |
| Gel size |
10cm W x 10cm L 10cm W x 8cm L |
16cm W x 14cm L 16cm W x 16cm L |
20cm W x 20cm L |
| Sample Capacity | 20 | 30 | 40 |
| Dimensions (cm) W x D x H | 15 x 13 x 13 | 20 x 13 x 18 | 25 x 13 x 23 |
| Glass Size(cm)WxL | 10 x 10 | 16 x 14 | 20 x 20 |
| Compatible w/ Owl Units | P8DS, P82 | P9DS | P10DS |
Attachment(s):
| File | Last Modified |
|---|---|
| 7217837 - Rev 0 - Multiple Gradient Caster, Models P7-CST, P1-CST and P2-CST - Operating and Maintenance Manual.pdf | December 15, 2021 |