7218313 Rev 0 - Sequencing Gel Casters Models SGC-1 and SGC-2 - Operating and Maintenance Manual
The document titled "7218313 - Rev 0 - Sequencing Gel Casters, Models SGC-1 and SGC-2 - Operating and Maintenance Manual" provides detailed information on the operation and maintenance of the Sequencing Gel Casters, models SGC-1 and SGC-2. It includes specifications, parts lists, safety precautions, cleaning instructions, and operating procedures for these models. The manual emphasizes easy and quick gel casting with minimal exposure to acrylamide.
Description:
Operating and maintenance manual
Environment:
Sequencing Gel Casters, models:
- SGC-1
- SGC-2
SGC-1:
- Gel Size: 20-42cm W x 48cm L
- Footprint: 21.6cm W x 71cm D (fully closed) x 12.2cm H
SGC-2:
- Gel Size: 20-42cm W x 65.5cm L
- Footprint: 21.2cm W x 102cm D (fully closed) x 12.2cm H
Attachment(s):
| File | Last Modified |
|---|---|
| 7218313 - Rev 0 - Sequencing Gel Casters, Models SGC-1 and SGC-2 - Operating and Maintenance Manual.pdf | December 15, 2021 |