Thermo Fisher Scientific
7219011 - Rev 0 - Horizontal System, Model A2-OK - Operating and Maintenance Manual
Description: 
Operating and maintenance manual
Environment:
Horizontal System Model A2-OK, specifications:
Gel size 7cmW x 8cmL
Running Buffer Volume 2.3L
Footprint 28cm W x 37.5cm L x 10cm H