LTM1596X1 - Owl Separation Systems Electrophoresis Power Supply OSP300Q - 120V and OSP300-2Q - 240V - User Manual
The manual for the Owl Separation Systems Electrophoresis Power Supply OSP300Q (120V) and OSP300-2Q (240V) includes comprehensive guidance on its setup, features, and operation. The OSP300 series provides constant voltage or current output for electrophoresis, supports up to three parallel electrophoresis cells, and delivers up to 75W of power. Key features include adjustable voltage and current, automatic control mode crossover, and a bright 3-digit display.
Description:
User manual
Environment:
Owl OSP300Q - 120V
Owl OSP300-2Q - 240V
Attachment(s):
| File | Last Modified |
|---|---|
| User_Manual - Owl OSP300Q and OSP300-2Q Electrophoresis Power Supply - Serial_Number Starting with 1596 - LT1596X1.pdf | June 27, 2025 |