LTM1596X2 - EC300XL Electrophoresis Power Supply - Operating Manual
The Thermo Scientific EC300XL Electrophoresis Power Supply Operating Manual provides detailed information on the setup, operation, safety, specifications, and maintenance of the EC300XL power supply. Key points include safety guidelines, specifications for AC and DC power inputs, operating procedures for constant voltage and current, and troubleshooting tips. It emphasizes the importance of using Thermo-approved apparatus and includes a warranty statement.
Description:
This manual describes the operation of the Thermo Scientific EC300 Electrophoresis power supply
Environment:
Thermo Scientific EC300 Electrophoresis Power Supply, specifications:
AC input Power
- 100-120 VAC, 50-60Hz, 100VA Max
Environmental
- Operating temperature: 0-40°C, 0-95% R.H. non-condensing
- Altitude: 2000m
- Overvoltage category II, IEC664
- Pollution degree 2, IEC664
DC output Power
- 10-300VDC, 75W Max
- 4-400mA, 75W Max
- Ripple: ± 1%
- Drift: ± 1%, after 30-minute warm-up
Attachment(s):
| File | Last Modified |
|---|---|
| LTM1596X2 - EC300XL Electrophoresis Power Supply - Operating Manual | January 19, 2022 |