OWL - All Horizontals Except for A5_B3 and D3 Models - Rewiring Procedure
To perform the rewiring procedure for OWL horizontal models (excluding A5, B3, and D3 models), follow these steps: 1. Clean the area and secure the banana plug with a nut. 2. Cut the wire protector from the near corner to the banana plug/nut. 3. Tack the platinum wire in the far corner using a soldering iron. 4. Tack the wire in the rear corner, adding 3-4 tacks. 5. Place the wire protector over the platinum wire. 6. Wrap the wire around the banana plug 2-3 times and cut it...
Description:
Instruction sheet for rewiring procedure
Environment:
OWL, all horizontals except for models:
- A5
- B3
- D3
Attachment(s):
| File | Last Modified |
|---|---|
| OWL, All Horizontals Except for A5, B3 and D3 Models - Rewiring Procedure.pdf | December 15, 2021 |