Thermo Fisher Scientific
The Opossum Horizontal System, Model A2-0K - Owner's Manual
Description: 
Owner's Manual
Environment:
Owl Separation Systems, The Opossum Horizontal Multiple Gel, Model A2-OK system
Specifications:
Gel size 7cmW x 8cmL (x6)
Running Buffer Volume 2.3L
Footprint 28cmW x 37.5cmL x 10cmH