The Opossum Horizontal System Model A2-0K - Owner's Manual
The Opossum Horizontal System Model A2-0K is designed for running multiple gels simultaneously, ideal for teaching labs or labs needing to run several gels at once. It can handle up to six minigels (7 cm x 8 cm) and screen up to 144 samples with optional combs. Key features include EasyCast design for gel casting without tape or plugs, and UV transmissible gel trays for photodocumentation. The system's footprint is 28 cm x 37.5 cm x 10 cm, with a running buffer volume of 2.3 liters.
Description:
Owner's Manual
Environment:
Owl Separation Systems, The Opossum Horizontal Multiple Gel, Model A2-OK system
Specifications:
| Gel size | 7cmW x 8cmL (x6) |
| Running Buffer Volume | 2.3L |
| Footprint | 28cmW x 37.5cmL x 10cmH |
Attachment(s):
| File | Last Modified |
|---|---|
| The Opossum Horizontal System, Model A2-0K - Owner's Manual.pdf | January 19, 2022 |