Thermo Fisher Scientific
LTM1595X2 - Thermo Scientific EC200XL - Electrophoresis Power Supply - Instruction Manual
The Thermo Scientific EC200XL Electrophoresis Power Supply Instruction Manual provides comprehensive guidance on operating, maintaining, and ensuring safety with the EC200XL power supply. It includes details on specifications, such as its ability to deliver 5-200V and 0.01-2.00A, with a maximum output of 200W. The manual emphasizes safety standards, usage instructions, and features like automatic control mode crossover and timed operations up to 99 hours and 59 minutes.
Description: 
EC-200XL Power Supply
Environment:
EC200xl, ec-200xl, ec200,
  • Was this article helpful?