Thermo Fisher Scientific
304233H01 - Rev C - Lindberg/Blue 1100C Box Furnace, BF51700 Series - Installation and Operation Manual
Description: 
Installation and operation manual
Environment:
Lindberg/Blue M 1100°C Box Furnace BF51700 Series:
BF51748Aa 120 VAC 50/60 Hz, 1 phase
BF51748Ca 208/240 VAC 50/60 Hz, 1 phase
BF51766Aa 120 VAC 50/60 Hz, 1 phase
BF51766Ca 208/240 VAC 50/60 Hz, 1 phase
BF51794Ca 208/240 VAC 50/60 Hz, 1 phase
BF51728Cb 208/240 VAC 50/60 Hz, 1 phase
aEquipped with power cord and plug: 208/240 VAC operation for "A" models, 120 VAC for "C" models
bNo power cord supplied; customer to complete wiring to power supply.