304233H01 - Rev C - Lindberg Blue 1100C Box Furnace BF51700 Series - Installation and Operation Manual
The Lindberg/Blue M 1100°C Box Furnace BF51700 Series installation and operation manual provides detailed instructions on installation, operation, and maintenance. Key sections include safety considerations, pre-installation steps, installation procedures, start-up process, operation using the UT150 controller, maintenance tasks, troubleshooting, replacement parts, wiring diagrams, and warranty information. The manual covers models BF51748A, BF51748C, BF51766A, BF51766C, BF51794C, and BF51728C.
Description:
Installation and operation manual
Environment:
Lindberg/Blue M 1100°C Box Furnace BF51700 Series:
| BF51748Aa | 120 VAC 50/60 Hz, 1 phase |
| BF51748Ca | 208/240 VAC 50/60 Hz, 1 phase |
| BF51766Aa | 120 VAC 50/60 Hz, 1 phase |
| BF51766Ca | 208/240 VAC 50/60 Hz, 1 phase |
| BF51794Ca | 208/240 VAC 50/60 Hz, 1 phase |
| BF51728Cb | 208/240 VAC 50/60 Hz, 1 phase |
aEquipped with power cord and plug: 208/240 VAC operation for "A" models, 120 VAC for "C" models
bNo power cord supplied; customer to complete wiring to power supply.
Attachment(s):
| File | Last Modified |
|---|---|
| 304233H01 - Rev C - Lindberg_Blue 1100C Box Furnace, BF51700 Series - Installation and Operation Manual.pdf | December 16, 2021 |