Thermo Fisher Scientific
304243H02 - Lindberg/Blue M Mini-Mite Tube Furnace - Installation and Operating Manual
Description: 
Installation and operating manual in French and English
Environment:
Lindberg/Blue M Mini-Mite TF55030 and TF55035 Series Moldatherm Tube Furnaces, models:
Models Electrical
TF55030 A-1 120 VAC, 50/60 Hz 800 W, 6.8 A
TF55030 COMA-1# 120 VAC, 50/60 Hz 800 W, 6.8 A
TF55030 C-1 208/240 VAC, 50/60 Hz 800 W, 3.3 A
TF55030 COMC-1# 208/240 VAC, 50/60 Hz 800 W, 3.3 A
TF55035 A-1 120 VAC, 50/60 Hz 800 W, 6.8 A
TF55035 COMA-1# 120 VAC, 50/60 Hz 800 W, 6.8 A
TF55035 C-1 208/240 VAC, 50/60 Hz 800 W, 3.3 A
TF55035 COMC-1# 208/240 VAC, 50/60 Hz 800 W, 3.3 A
# Unit includes male and female RS 485 Digital Communications DB-9 ports