304243H02 RevC and RevD - Lindberg Blue M Mini-Mite Tube Furnace - Installation and Operating Manual
The Lindberg/Blue M Mini-Mite Tube Furnace installation and operating manual provides instructions for models TF55030 and TF55035. The manual includes electrical specifications, start-up procedures, safety precautions, and operational guidelines. Key safety measures include wearing protective gear and ensuring the furnace door is closed to activate the power. The manual is available in French and English.
Description:
Installation and operating manual in French and English
Environment:
Lindberg/Blue M Mini-Mite TF55030 and TF55035 Series Moldatherm Tube Furnaces, models:
| Models | Electrical |
| TF55030 A-1 | 120 VAC, 50/60 Hz 800 W, 6.8 A |
| TF55030 COMA-1# | 120 VAC, 50/60 Hz 800 W, 6.8 A |
| TF55030 C-1 | 208/240 VAC, 50/60 Hz 800 W, 3.3 A |
| TF55030 COMC-1# | 208/240 VAC, 50/60 Hz 800 W, 3.3 A |
| TF55035 A-1 | 120 VAC, 50/60 Hz 800 W, 6.8 A |
| TF55035 COMA-1# | 120 VAC, 50/60 Hz 800 W, 6.8 A |
| TF55035 C-1 | 208/240 VAC, 50/60 Hz 800 W, 3.3 A |
| TF55035 COMC-1# | 208/240 VAC, 50/60 Hz 800 W, 3.3 A |
# Unit includes male and female RS 485 Digital Communications DB-9 ports