305378H01 Rev H - Lindberg Blue 1700C Box Furnace - User Manual
The "305378H01 - Lindberg/Blue 1700C Box Furnace - User Manual" provides detailed guidance on the BF51634 and BF51664 series box furnaces. It covers specifications, installation, operation, and maintenance for models such as BF51634C, BF51634BC, BF51664C, and others. These furnaces operate at 208/240 volts, with wattages of 5900 (BF51634 series) and 7100 (BF51664 series).
Description:
User Manual
Environment:
BF51634/BF51664 Series Box Furnace, models:
| Model | Watts | Voltage (50/60 Hz) |
| BF51634C | 5900 | 208/240 |
| BF51634BC | 5900 | 208/240 |
| BF51634PCOMC | 5900 | 208/240 |
| BF51634PBCOMC | 5900 | 208/240 |
| BF51664C | 7100 | 208/240 |
| BF51664BC | 7100 | 208/240 |
| BF51664PC | 7100 | 208/240 |
| BF51664PBC | 7100 | 208/240 |
| BF51664PCOMC | 7100 | 208/240 |
| BF51664PBCOMC | 7100 | 208/240 |
Attachment(s):
| File | Last Modified |
|---|---|
| 305378H01 - Rev H - LindbergBlue 1700C Box Furnace - User Manual.pdf | December 17, 2021 |
| 305378H01 - Rev G - Lindberg_Blue 1700C Box Furnace - User Manual.pdf | December 16, 2021 |