Thermo Fisher Scientific
305378H02 - Rev C - Lindberg/Blue M 1700C Box Furnaces, BF51634/ BF51664 Series - Installation and Operational Manual
Description: 
Installation and operational manual
Environment:
 Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces, BF51634/ BF51664 Series, models:
Model Watts Volt, Freq., Current
BF51634PC-1 5900 208/240 V, 50/60 Hz, 23 A
BF51634PCOMC-1 5900 208/240 V, 50/60 Hz, 23 A
BF51634PBC-1 5900 208/240 V, 50/60 Hz, 23 A
BF51634PBCOMC-1 5900 208/240 V, 50/60 Hz, 23 A
BF51664PC-1 7100 208/240 V, 50/60 Hz, 30 A
BF51664PCOMC-1 7100 208/240 V, 50/60 Hz, 30 A
BF51664PBC-1 7100 208/240 V, 50/60 Hz, 30 A
BF51664PBCOMC-1 7100 208/240 V, 50/60 Hz, 30 A