305378H02 - Rev C - Lindberg BlueM 1700C Box Furnaces_BF51634_BF51664 Series - Installation and Operational Manual
The manual for Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces, BF51634/BF51664 Series, provides comprehensive instructions for installation and operation. It includes specifications for models BF51634PC-1, BF51634PCOMC-1, BF51634PBC-1, BF51634PBCOMC-1, BF51664PC-1, BF51664PCOMC-1, BF51664PBC-1, and BF51664PBCOMC-1.
Description:
Installation and operational manual
Environment:
Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces, BF51634/ BF51664 Series, models:
| Model | Watts | Volt, Freq., Current |
| BF51634PC-1 | 5900 | 208/240 V, 50/60 Hz, 23 A |
| BF51634PCOMC-1 | 5900 | 208/240 V, 50/60 Hz, 23 A |
| BF51634PBC-1 | 5900 | 208/240 V, 50/60 Hz, 23 A |
| BF51634PBCOMC-1 | 5900 | 208/240 V, 50/60 Hz, 23 A |
| BF51664PC-1 | 7100 | 208/240 V, 50/60 Hz, 30 A |
| BF51664PCOMC-1 | 7100 | 208/240 V, 50/60 Hz, 30 A |
| BF51664PBC-1 | 7100 | 208/240 V, 50/60 Hz, 30 A |
| BF51664PBCOMC-1 | 7100 | 208/240 V, 50/60 Hz, 30 A |
Attachment(s):
| File | Last Modified |
|---|---|
| 305378H02 - Rev C - Lindberg_Blue M 1700C Box Furnaces, BF51634_ BF51664 Series - Installation and Operational Manual.pdf | December 17, 2021 |