Thermo Fisher Scientific
305395H02 - Rev B - Thermo Scientific Lindberg/Blue M 1700C Box Furnaces Independent Control Consoles - Installation and Operational Manual
Description: 
Installation and operational manual
Environment:
Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces Independent Control Consoles, furnace models:
Model Watts Electrical (Heater)
BF51314C 3500 58V 50/60Hz 60A
BF51524C 5000 100V 50/60Hz 50A
Control Console models:
For Model BF51314C:
Model Electrical
CC59246PCOMC-1 208/240V, 50/60Hz 30 A
CC59246PBCOMC-1 208/240V, 50/60Hz 30 A
For Model BF51524C:
Model Electrical
CC59256PCOMC-1 208/240V, 50/60Hz 30 A
CC59256PBCOMC-1 208/240V, 50/60Hz 30 A
CC59256PCM2CTC-1 208/240V, 50/60Hz 30 A