305395H02 - Rev B - Thermo Scientific Lindberg BlueM 1700C Box Furnaces Independent Control Consoles - Installation and Operational Manual
The "305395H02 - Rev B - Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces Independent Control Consoles - Installation and Operational Manual" provides detailed installation and operational instructions for the Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces and their independent control consoles. It includes comprehensive information on installation procedures, operational guidelines, and maintenance instructions for furnace models BF51314C and BF51524C, and their respective control c
Description:
Installation and operational manual
Environment:
Thermo Scientific Lindberg/Blue M 1700°C Box Furnaces Independent Control Consoles, furnace models:
| Model | Watts | Electrical (Heater) |
| BF51314C | 3500 | 58V 50/60Hz 60A |
| BF51524C | 5000 | 100V 50/60Hz 50A |
Control Console models:
| For Model BF51314C: | |
| Model | Electrical |
| CC59246PCOMC-1 | 208/240V, 50/60Hz 30 A |
| CC59246PBCOMC-1 | 208/240V, 50/60Hz 30 A |
| For Model BF51524C: | |
| Model | Electrical |
| CC59256PCOMC-1 | 208/240V, 50/60Hz 30 A |
| CC59256PBCOMC-1 | 208/240V, 50/60Hz 30 A |
| CC59256PCM2CTC-1 | 208/240V, 50/60Hz 30 A |
Attachment(s):
| File | Last Modified |
|---|---|
| 305395H02 - Rev B - Thermo Scientific Lindberg_Blue M 1700C Box Furnaces Independent Control Consoles - Installation and Operational Manual.pdf | December 17, 2021 |