305432H02 - Rev C - Thermo Scientific Lindberg BlueM 1200C Box Furnaces BF 51842 Series - Installation and Operational Manual
The document "305432H02 - Rev C - Thermo Scientific Lindberg/Blue M 1200°C Box Furnaces, BF 51842 Series - Installation and Operational Manual" provides comprehensive guidelines for installing and operating the BF 51842 Series models. It includes detailed instructions on setup, usage, and maintenance of the furnaces. The manual is essential for ensuring proper functionality and safety during the operation of these high-temperature furnaces.
Description:
Installation and operational manual
Environment:
Thermo Scientific Lindberg/Blue M LGO 1200°C Box Furnaces BF 51842 Series, models:
| Model | Watts | Volt/Freq/Current |
| BF51842C-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842COMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842BC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PBC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PBFMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PBCOMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PBFMCOMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PFMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PCOMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
| BF51842PFMCOMC-1 | 5800 | 208/240 V 50/60 Hz 25 A |
Attachment(s):
| File | Last Modified |
|---|---|
| 305432H02 - Rev C - Thermo Scientific Lindberg_Blue M 1200C Box Furnaces, BF 51842 Series - Installation and Operational Manual.pdf | December 17, 2021 |