Thermo Fisher Scientific
39129H01-RC - Lindberg/Blue Moldatherm Box Furnaces, BF51600, BF51700, and BF51800 Series - Installation and Operation Manual
Description: 
Installation and operation manual
Environment:
Lindberg/Blue Moldatherm, BF51600, BF51700, and BF51800 Series Moldatherm Box Furnaces, models:
Model Watts Voltage
BF51648Aa 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51648Ca 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51748Aa 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51748Ca 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51848Aa 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51848Ca 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51666Aa 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51666Ca 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51766Aa 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51766Ca 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51866Aa 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51866Ca 1800 120/208/240 VAC 50/60 HZ, 1 Phase
BF51694Cb 3500 208/240 VAC 50/60 HZ, 1 Phase
BF51794Cb 3500 208/240 VAC 50/60 HZ, 1 Phase
BF51894Cb 3500 208/240 VAC 50/60 HZ, 1 Phase
BF51628C 5600 208/240 VAC 50/60 HZ, 1 Phase
BF51728C 5600 208/240 VAC 50/60 HZ, 1 Phase
BF51828C 5600 208/240 VAC 50/60 HZ, 1 Phase
aVoltage Code: A - 120V Model; C - 208/240V Model. “A" models are equipped with power cord and plug for 120 VAC operation. “C" models are equipped with power cord and plug for 208/240 VAC operation.
bEquipped with power cord and plug for 240 VAC operation. q