39129H01-RC - Lindberg Blue Moldatherm Box Furnaces_BF51600_BF51700 and BF51800 Series - Installation and Operation Manual
The manual for Lindberg/Blue Moldatherm Box Furnaces, BF51600, BF51700, and BF51800 series includes detailed guidance on installation, operation, maintenance, and troubleshooting. Key sections cover safety guidelines, unpacking and operating conditions, location and wiring requirements, start-up procedures, operation instructions for different controllers, maintenance tips, troubleshooting common issues, and replacement parts with wiring diagrams. The document ensures proper handling and efficie
Description:
Installation and operation manual
Environment:
Lindberg/Blue Moldatherm, BF51600, BF51700, and BF51800 Series Moldatherm Box Furnaces, models:
| Model | Watts | Voltage | |
| BF51648Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51648Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51748Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51748Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51848Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51848Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51666Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51666Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51766Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51766Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51866Aa | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51866Ca | 1800 | 120/208/240 VAC 50/60 HZ, 1 Phase | |
| BF51694Cb | 3500 | 208/240 VAC 50/60 HZ, 1 Phase | |
| BF51794Cb | 3500 | 208/240 VAC 50/60 HZ, 1 Phase | |
| BF51894Cb | 3500 | 208/240 VAC 50/60 HZ, 1 Phase | |
| BF51628C | 5600 | 208/240 VAC 50/60 HZ, 1 Phase | |
| BF51728C | 5600 | 208/240 VAC 50/60 HZ, 1 Phase | |
| BF51828C | 5600 | 208/240 VAC 50/60 HZ, 1 Phase | |
aVoltage Code: A - 120V Model; C - 208/240V Model. “A" models are equipped with power cord and plug for 120 VAC operation. “C" models are equipped with power cord and plug for 208/240 VAC operation.
bEquipped with power cord and plug for 240 VAC operation. q
Attachment(s):
| File | Last Modified |
|---|---|
| 39129H01-RC - Lindberg_Blue Moldatherm Box Furnaces, BF51600, BF51700, and BF51800 Series - Installation and Operation Manual.pdf | December 17, 2021 |