39129H18 - Lindberg Blue 1100C Box Furnace BF51800 Series - Installation and Operation Manual
The "39129H18 - Lindberg/Blue 1100°C Box Furnace, BF51800 Series - Installation and Operation Manual" provides detailed instructions for setting up, operating, maintaining, and troubleshooting the Lindberg/Blue 1100°C Box Furnace models. It includes guidelines on safety, pre-installation, wiring for different voltages, furnace startup, controller operation, component replacement, and resolving common issues. The manual also contains wiring diagrams and a list of replacement parts.
Description:
Installation and operation manual
Environment:
1100°C Box Furnace, BF51800 Series, models:
| Model | Watts | Voltage |
| BF51848A | 1800 | 120/VAC 50/60 Hz |
| BF51848C | 1800 | 208/240 VAC 50/60 Hz |
| BF51866A | 1800 | 120/VAC 50/60 Hz |
| BF51866C | 1800 | 208/240 VAC 50/60 Hz |
| BF51894C | 3500 | 208/240 VAC 50/60 Hz |
| BF51828C | 5600 | 208/240 VAC 50/60 Hz |
Attachment(s):
| File | Last Modified |
|---|---|
| Operation Manual 330985H01_revD_BF51700 BF51800 Uplift Nov 2022.pdf | November 21, 2023 |
| 39129H18 - Lindberg_Blue 1100C Box Furnace, BF51800 Series - Installation and Operation Manual.pdf | December 17, 2021 |