F-144 - Thermolyne Electrical Furnace Type F1400 - User Manual and Parts List
The Thermolyne Electrical Furnace Type F1400 user manual and parts list provide detailed information about the furnace, including model specifications, features, and a comprehensive parts list. The furnace is designed for laboratory and plant use, featuring a sturdy steel case, embedded nickel-chromium heating elements, a pyrometer for temperature indication, and a built-in controller for stepless temperature control. It operates on alternating current and includes a 3-wire cord and plug.
Description:
User manual and parts list
Environment:
Thermolyne electrical furnace type F1400 for serial numbers starting With 14
| Model Number | Volts 50/60 cycle | Amps | Max watts |
| F-A1410M | 230 | 6. 1 | 1400 |
| F-A1415M | 115 | 12.2 | 1400 |
Attachment(s):
| File | Last Modified |
|---|---|
| F-144 - Thermolyne Electrical Furnace Type F1400 - User Manual and Parts List.pdf | February 01, 2022 |