Thermo Fisher Scientific
L-83102-51442 - Rev A - Thermo Scientific Lindberg/Blue M 1200C/1500C Box Furnaces BF51000 Series - Installation and Operational Manual
Description: 
These operating instructions describe 1200°C/1500°C Box Furnaces.
Environment:
Thermo Scientific Lindberg/Blue M 1200°C/1500°C Box Furnaces BF51000 Series, models:
Model Watts Electrical
BF51442C 1200°C 208/240 V 50/60 HZ 21 A
BF51542C 1200°C 208/240 V 50/60 HZ 26 A
BF51333C 1500°C 208/240 V 50/60 HZ 25 A