L-87118-55122-1 - Lindberg/Blue HTF55000 Series Hinged Tube Furnaces - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Lindberg/Blue HTF55000 Series Moldatherm Hinged Tube Furnaces, models:
Furnace Model # | AC Volts Single Phase 50/60 HZ | Power (Watts) |
Single Zone | ||
55122 | 120 | 1330 |
55322 | 120/240 | 2675 |
55332 | 240 | 4240 |
55342 | 240 | 5440 |
55642 | 240 | 8710 |
55662 | 240 | 12195 |
Three Zone | ||
55347 | 240 | 5355 |
55367 | 240 | 8025 |
55647 | 240 | 7840 |
55667 | 240 | 11760 |
Attachment(s):
File | Last Modified |
---|---|
L-87118-55122-1 - Lindberg_Blue HTF55000 Series Hinged Tube Furnaces - Installation and Operation Manual.pdf | December 17, 2021 |