L-87118-55122-1 - Lindberg Blue HTF55000 Series Hinged Tube Furnaces - Installation and Operation Manual
The Lindberg/Blue HTF55000 Series Hinged Tube Furnaces Installation and Operation Manual provides comprehensive guidelines for installing, operating, and maintaining these furnaces. Key points include placement on a level surface, wiring instructions, and safety precautions regarding exposure to refractory ceramic fiber and silica dust. Electrical specifications for various models are detailed, and initial start-up involves operating the furnace at 550°C for one hour to ensure proper functionali
Description:
Installation and operation manual
Environment:
Lindberg/Blue HTF55000 Series Moldatherm Hinged Tube Furnaces, models:
| Furnace Model # | AC Volts Single Phase 50/60 HZ | Power (Watts) |
| Single Zone | ||
| 55122 | 120 | 1330 |
| 55322 | 120/240 | 2675 |
| 55332 | 240 | 4240 |
| 55342 | 240 | 5440 |
| 55642 | 240 | 8710 |
| 55662 | 240 | 12195 |
| Three Zone | ||
| 55347 | 240 | 5355 |
| 55367 | 240 | 8025 |
| 55647 | 240 | 7840 |
| 55667 | 240 | 11760 |
Attachment(s):
| File | Last Modified |
|---|---|
| L-87118-55122-1 - Lindberg_Blue HTF55000 Series Hinged Tube Furnaces - Installation and Operation Manual.pdf | December 17, 2021 |