Thermo Fisher Scientific
LT1087X2 - Rev G - Barnstead Thermolyne NCAT Asphalt Content Tester, Series 1087 - Operation Manual and Parts List
Description: 
Operation manual and parts List
Environment:
 Barnstead  | Thermolyne NCAT Asphalt Content Tester, Series 1087, models:
Model No. Phase Voltage Watts Amps
F85930 1 220-240 4879/6379 20/27
F585938 1 208 5757 28
F85930-33 1 220-240 4879/6379 20/27
45-3790/01 1 220-240 4879/6379 20/27
45-3790/07 1 208 5757 28