LT1087X2 - Rev G - Barnstead Thermolyne NCAT Asphalt Content Tester Series 1087 - Operation Manual and Parts List
The "LT1087X2 - Rev G - Barnstead Thermolyne NCAT Asphalt Content Tester, Series 1087 - Operation Manual and Parts List" provides details on the operation and parts list for the NCAT Asphalt Content Tester, Series 1087. It includes safety information, warnings, general usage instructions, software updates, theory of operation, and general specifications. The manual emphasizes the importance of proper installation by a qualified electrician and warns against using the product for cleaning glasswa
Description:
Operation manual and parts List
Environment:
Barnstead | Thermolyne NCAT Asphalt Content Tester, Series 1087, models:
| Model No. | Phase | Voltage | Watts | Amps |
| F85930 | 1 | 220-240 | 4879/6379 | 20/27 |
| F585938 | 1 | 208 | 5757 | 28 |
| F85930-33 | 1 | 220-240 | 4879/6379 | 20/27 |
| 45-3790/01 | 1 | 220-240 | 4879/6379 | 20/27 |
| 45-3790/07 | 1 | 208 | 5757 | 28 |
Attachment(s):
| File | Last Modified |
|---|---|
| LT1087X2 - Rev G - Barnstead Thermolyne NCAT Asphalt Content Tester, Series 1087 - Operation Manual and Parts List.pdf | December 16, 2021 |