LT1170X1 - Thermo Scientific Thermolyne Furnace Benchtop Industrial, Type FD1500M - Installation and Operation Manual
Description:
Installation and operation manual
Note: Rev G has updated 3216c and 3216p controller.
Environment:
Thermo Scientific Thermolyne Furnace Benchtop Industrial, Type FD1500M, model numbers:
Model Number: | FD1540M, FD1530M | FD1540M-33 FD1530M-33 | FD1545M FD1535M |
Volts: | 240 | 220-240 | 120 |
Amps: | 9.3 | 6.5 | 18.6 |
Watts: | 2230 | 1560 | 2230 |
Freq: | 50/60 | 50/60 | 50/60 |
Phase: | 1 | 1 | 1 |