LT1170X1_Rev G - Thermo Scientific Thermolyne Furnace Benchtop Industrial_Type FD1500M - Installation and Operation Manual
The article titled "Thermo Scientific Thermolyne Furnace Benchtop Industrial, Type FD1500M - Installation and Operation Manual" addresses the Rev G update for the 3216c and 3216p controllers. The environment includes model numbers FD1540M, FD1530M, FD1540M-33, FD1530M-33, FD1545M, and FD1535M. The resolution involves following the updated installation and operation procedures for the 3216c and 3216p controllers as specified in Rev G. The cause of the issue is an update in the controller specific
Description:
Installation and operation manual
Note: Rev G has updated 3216c and 3216p controller.
Environment:
Thermo Scientific Thermolyne Furnace Benchtop Industrial, Type FD1500M, model numbers:
| Model Number: | FD1540M, FD1530M | FD1540M-33 FD1530M-33 | FD1545M FD1535M |
| Volts: | 240 | 220-240 | 120 |
| Amps: | 9.3 | 6.5 | 18.6 |
| Watts: | 2230 | 1560 | 2230 |
| Freq: | 50/60 | 50/60 | 50/60 |
| Phase: | 1 | 1 | 1 |