LT1256X1 - Thermo Scientific Thermolyne Furnace, Small Benchtop Muffle, Type FB1300 and FB1400 - Installation and Operation Manual
Description:
Installation and operation manual
Environment:
Thermo Scientific Thermolyne Furnace, Small Benchtop Muffle, Type FB1300 and FB1400
FB1300 models:
Model # | Volts | Amps | Watts | Phase | Frequency |
FB1310M | 220-240 | 4.4 | 1060 | 1 | 50/60 |
FB1310M-26 | 220-240 | 4.4 | 1060 | 1 | 50/60 |
FB1310M-33 | 220-240 | 4.4 | 1060 | 1 | 50/60 |
FB1310M-33CN | 220-240 | 4.4 | 1060 | 1 | 50/60 |
FB1310M-33CH | 220-240 | 4.4 | 1060 | 1 | 50/60 |
FB1310M-33CK | 220-240 | 4.4 | 1060 | 1 | 50/60 |
FB1314M | 100 | 10.6 | 1060 | 1 | 50/60 |
FB1315M | 120 | 8.9 | 1060 | 1 | 50/60 |
FB1318M | 208 | 5.1 | 1060 | 1 | 50/60 |
FB1400 models:
Model # | Volts | Amps | Watts | Phase | Frequency |
FB1410M | 220-240 | 6.3 | 1520 | 1 | 50/60 |
FB1410M-26 | 220-240 | 6.3 | 1520 | 1 | 50/60 |
FB1410M-33 | 220-240 | 6.3 | 1520 | 1 | 50/60 |
FB1410M-33CN | 220-240 | 6.3 | 1520 | 1 | 50/60 |
FB1410M-33CH | 220-240 | 6.3 | 1520 | 1 | 50/60 |
FB1410M-33UK | 220-240 | 6.3 | 1520 | 1 | 50/60 |
FB1414M | 100 | 14.5 | 1520 | 1 | 50/60 |
FB1415M | 120 | 12 | 1450 | 1 | 50/60 |
FB1418M | 208 | 7.3 | 1520 | 1 | 50/60 |